RIE156330139 Rieke Metals, Inc.

2,2-Dimethyl-4-oxo-4-(3,4,5-trimethoxyphenyl)butyric acid

  • MDL number MFCD09801853
  • Molecular Weight : 296.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7287

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7287
Availability details for
Quantity
  • Availability
  • Add to Cart