RIE156330140 Rieke Metals, Inc.

3-Methyl-5-oxo-5-(3,4,5-trimethoxyphenyl)valeric acid

  • MDL number MFCD09801854
  • Cas Number : 101499-91-6
  • Molecular Weight : 296.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7288

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7288
Availability details for
Quantity
  • Availability
  • Add to Cart