RIE156330142 Rieke Metals, Inc.

3-Methyl-5-(3-methyl-2-thienyl)-5-oxovaleric acid

  • MDL number MFCD09801856
  • Molecular Weight : 226.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7290

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7290
Availability details for
Quantity
  • Availability
  • Add to Cart