RIE156330147 Rieke Metals, Inc.

3-Methyl-5-[3,4-(methylenedioxy)phenyl]-5-oxovaleric acid

  • MDL number MFCD09801860
  • Molecular Weight : 250.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7296

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7296
Availability details for
Quantity
  • Availability
  • Add to Cart