RIE156330154 Rieke Metals, Inc.

5-(2,5-Dimethylphenyl)-3-methyl-5-oxovaleric acid

  • MDL number MFCD09801867
  • Cas Number : 92864-23-8
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7304

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7304
Availability details for
Quantity
  • Availability
  • Add to Cart