RIE156330157 Rieke Metals, Inc.

2,2-Dimethyl-4-(3,4-dimethylphenyl)-4-oxobutyric acid

  • MDL number MFCD09801870
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7307

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7307
Availability details for
Quantity
  • Availability
  • Add to Cart