RIE156330158 Rieke Metals, Inc.

5-(3,4-Dimethylphenyl)-3-methyl-5-oxovaleric acid

  • MDL number MFCD09801871
  • Cas Number : 92864-24-9
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7308

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7308
Availability details for
Quantity
  • Availability
  • Add to Cart