RIE156330159 Rieke Metals, Inc.

4-(2,4-Dimethoxyphenyl)-2,2-dimethyl-4-oxobutyric acid

  • MDL number MFCD09801872
  • Molecular Weight : 266.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7310

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7310
Availability details for
Quantity
  • Availability
  • Add to Cart