RIE156330165 Rieke Metals, Inc.

5-(3,5-Dimethoxyphenyl)-3-methyl-5-oxovaleric acid

  • MDL number MFCD09801878
  • Molecular Weight : 266.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7317

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7317
Availability details for
Quantity
  • Availability
  • Add to Cart