RIE156330171 Rieke Metals, Inc.

5-(4-Fluoro-3-methylphenyl)-3-methyl-5-oxovaleric acid

  • MDL number MFCD09801884
  • Molecular Weight : 238.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7323

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7323
Availability details for
Quantity
  • Availability
  • Add to Cart