RIE156330177 Rieke Metals, Inc.

5-(5-Methoxy-2-methylphenyl)-3-methyl-5-oxovaleric acid

  • MDL number MFCD09801890
  • Molecular Weight : 250.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7329

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7329
Availability details for
Quantity
  • Availability
  • Add to Cart