RIE156330180 Rieke Metals, Inc.

5-(5-Fluoro-2-methoxyphenyl)-3-methyl-5-oxovaleric acid

  • MDL number MFCD09801893
  • Molecular Weight : 254.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7333

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7333
Availability details for
Quantity
  • Availability
  • Add to Cart