RIE156330182 Rieke Metals, Inc.

5-(4-Chloro-2-methylphenyl)-3-methyl-5-oxovaleric acid

  • MDL number MFCD09801895
  • Molecular Weight : 254.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7335

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7335
Availability details for
Quantity
  • Availability
  • Add to Cart