RIE156330184 Rieke Metals, Inc.

5-(3-Chloro-4-methylphenyl)-3-methyl-5-oxovaleric acid

  • MDL number MFCD09801897
  • Molecular Weight : 254.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7337

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7337
Availability details for
Quantity
  • Availability
  • Add to Cart