RIE156330186 Rieke Metals, Inc.

4-(2-Chlorophenyl)-4-oxobutyric acid

  • MDL number MFCD03844205
  • Cas Number : 106263-50-7
  • Molecular Weight : 212.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7346B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7346B
Availability details for
Quantity
  • Availability
  • Add to Cart