RIE156330188 Rieke Metals, Inc.

6-(2-chlorophenyl)-6-oxohexanoic acid

  • MDL number MFCD03844207
  • Cas Number : 898792-61-5
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7346D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7346D
Availability details for
Quantity
  • Availability
  • Add to Cart