RIE156330190 Rieke Metals, Inc.

8-(2-chlorophenyl)-8-oxooctanoic acid

  • MDL number MFCD03844209
  • Cas Number : 898792-65-9
  • Molecular Weight : 268.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7346F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7346F
Availability details for
Quantity
  • Availability
  • Add to Cart