RIE156330192 Rieke Metals, Inc.

Ethyl 6-(4-bromophenyl)-6-oxohexanoate

  • MDL number MFCD00672070
  • Cas Number : 412022-61-8
  • Molecular Weight : 313.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7501D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7501D
Availability details for
Quantity
  • Availability
  • Add to Cart