RIE156330198 Rieke Metals, Inc.

ethyl 8-(3-bromophenyl)-8-oxooctanoate

  • MDL number MFCD01320181
  • Cas Number : 898792-73-9
  • Molecular Weight : 341.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7502F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7502F
Availability details for
Quantity
  • Availability
  • Add to Cart