RIE156330199 Rieke Metals, Inc.

Ethyl 5-(4-fluorophenyl)-5-oxovalerate

  • MDL number MFCD01311503
  • Cas Number : 342636-36-6
  • Molecular Weight : 238.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7503C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7503C
Availability details for
Quantity
  • Availability
  • Add to Cart