RIE156330203 Rieke Metals, Inc.

Ethyl 6-(4-chlorophenyl)-6-oxohexanoate

  • MDL number MFCD01320185
  • Cas Number : 54029-05-9
  • Molecular Weight : 268.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7504D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7504D
Availability details for
Quantity
  • Availability
  • Add to Cart