RIE156330204 Rieke Metals, Inc.

ethyl 4-(2,5-dimethylphenyl)-4-oxobutyrate

  • MDL number MFCD01320188
  • Cas Number : 30005-21-1
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7505B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7505B
Availability details for
Quantity
  • Availability
  • Add to Cart