RIE156330206 Rieke Metals, Inc.

ethyl 6-(2,5-dimethylphenyl)-6-oxohexanoate

  • MDL number MFCD01320190
  • Cas Number : 898792-82-0
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7505D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7505D
Availability details for
Quantity
  • Availability
  • Add to Cart