RIE156330207 Rieke Metals, Inc.

ethyl 7-(2,5-dimethylphenyl)-7-oxoheptanoate

  • MDL number MFCD01320191
  • Cas Number : 898792-85-3
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7505E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7505E
Availability details for
Quantity
  • Availability
  • Add to Cart