RIE156330209 Rieke Metals, Inc.

ethyl 4-(3,5-ditrifluoromethylphenyl)-4-oxobutyrate

  • MDL number MFCD01311605
  • Cas Number : 898792-91-1
  • Molecular Weight : 342.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7508B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7508B
Availability details for
Quantity
  • Availability
  • Add to Cart