RIE156330211 Rieke Metals, Inc.

ethyl 6-(3,5-ditrifluoromethylphenyl)-6-oxohexanoate

  • MDL number MFCD01311607
  • Cas Number : 898792-97-7
  • Molecular Weight : 370.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7508D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7508D
Availability details for
Quantity
  • Availability
  • Add to Cart