RIE156330216 Rieke Metals, Inc.

ethyl 5-(3,4-dimethylphenyl)-5-oxovalerate

  • MDL number MFCD01320197
  • Cas Number : 898793-06-1
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7512C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7512C
Availability details for
Quantity
  • Availability
  • Add to Cart