RIE156330219 Rieke Metals, Inc.

ethyl 8-(3,4-dimethylphenyl)-8-oxooctanoate

  • MDL number MFCD01320200
  • Cas Number : 898793-15-2
  • Molecular Weight : 290.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7512F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7512F
Availability details for
Quantity
  • Availability
  • Add to Cart