RIE156330220 Rieke Metals, Inc.

Ethyl 4-(3,5-dimethyl-4-methoxyphenyl)-4-oxobutyrate

  • MDL number MFCD01320201
  • Cas Number : 898793-18-5
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7513B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7513B
Availability details for
Quantity
  • Availability
  • Add to Cart