RIE156330223 Rieke Metals, Inc.

Ethyl 7-(3,5-dimethyl-4-methoxyphenyl)-7-oxoheptanoate

  • MDL number MFCD01320204
  • Cas Number : 898751-10-5
  • Molecular Weight : 306.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7513E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7513E
Availability details for
Quantity
  • Availability
  • Add to Cart