RIE156330226 Rieke Metals, Inc.

Ethyl 5-(2-bromophenyl)-5-oxovalerate

  • MDL number MFCD00672061
  • Cas Number : 898751-22-9
  • Molecular Weight : 299.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7514C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7514C
Availability details for
Quantity
  • Availability
  • Add to Cart