RIE156330228 Rieke Metals, Inc.

ethyl 7-(2-bromophenyl)-7-oxoheptanoate

  • MDL number MFCD00672063
  • Cas Number : 898751-30-9
  • Molecular Weight : 327.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7514E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7514E
Availability details for
Quantity
  • Availability
  • Add to Cart