RIE156330229 Rieke Metals, Inc.

ethyl 8-(2-bromophenyl)-8-oxooctanoate

  • MDL number MFCD01320206
  • Cas Number : 898751-34-3
  • Molecular Weight : 341.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7514F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7514F
Availability details for
Quantity
  • Availability
  • Add to Cart