RIE156330231 Rieke Metals, Inc.

Ethyl 4-(2-methylphenyl)-4-oxobutyrate

  • MDL number MFCD01320208
  • Cas Number : 79034-93-8
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7523B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7523B
Availability details for
Quantity
  • Availability
  • Add to Cart