RIE156330232 Rieke Metals, Inc.

ethyl 5-(2-methylphenyl)-5-oxovalerate

  • MDL number MFCD01320209
  • Cas Number : 100972-13-2
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7523C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7523C
Availability details for
Quantity
  • Availability
  • Add to Cart