RIE156330233 Rieke Metals, Inc.

ethyl 6-(2-methylphenyl)-6-oxohexanoate

  • MDL number MFCD01320210
  • Cas Number : 898751-38-7
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7523D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7523D
Availability details for
Quantity
  • Availability
  • Add to Cart