RIE156330234 Rieke Metals, Inc.

ethyl 7-(2-methylphenyl)-7-oxoheptanoate

  • MDL number MFCD01320211
  • Cas Number : 122115-55-3
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7523E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7523E
Availability details for
Quantity
  • Availability
  • Add to Cart