RIE156330236 Rieke Metals, Inc.

Ethyl 4-(3-methylphenyl)-4-oxobutyrate

  • MDL number MFCD01320213
  • Cas Number : 73931-65-4
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7527B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7527B
Availability details for
Quantity
  • Availability
  • Add to Cart