RIE156330238 Rieke Metals, Inc.

Ethyl 6-(3-methylphenyl)-6-oxohexanoate

  • MDL number MFCD01320215
  • Cas Number : 898751-50-3
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7527D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7527D
Availability details for
Quantity
  • Availability
  • Add to Cart