RIE156330242 Rieke Metals, Inc.

ethyl 6-(4-methylphenyl)-6-oxohexanoate

  • MDL number MFCD01320219
  • Cas Number : 100847-87-8
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7531D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7531D
Availability details for
Quantity
  • Availability
  • Add to Cart