RIE156330244 Rieke Metals, Inc.

Ethyl 8-(4-methylphenyl)-8-oxooctanoate

  • MDL number MFCD01320221
  • Cas Number : 898751-62-7
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7531F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7531F
Availability details for
Quantity
  • Availability
  • Add to Cart