RIE156330245 Rieke Metals, Inc.

ethyl 2,2-dimethyl-4-(3,5-dimethylphenyl)-4-oxobutyrate

  • MDL number MFCD01320222
  • Cas Number : 898751-66-1
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7535

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7535
Availability details for
Quantity
  • Availability
  • Add to Cart