RIE156330246 Rieke Metals, Inc.

ethyl 4-(3,5-dimethylphenyl)-4-oxobutyrate

  • MDL number MFCD01320223
  • Cas Number : 898751-69-4
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7536B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7536B
Availability details for
Quantity
  • Availability
  • Add to Cart