RIE156330247 Rieke Metals, Inc.

ethyl 5-(3,5-dimethylphenyl)-5-oxovalerate

  • MDL number MFCD01320224
  • Cas Number : 898751-72-9
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7536C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7536C
Availability details for
Quantity
  • Availability
  • Add to Cart