RIE156330251 Rieke Metals, Inc.

Ethyl 4-(3,5-dichlorophenyl)-4-oxobutyrate

  • MDL number MFCD01320228
  • Cas Number : 898751-84-3
  • Molecular Weight : 275.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7538B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7538B
Availability details for
Quantity
  • Availability
  • Add to Cart