RIE156330255 Rieke Metals, Inc.

ethyl 8-(3,5-dichlorophenyl)-8-oxooctanoate

  • MDL number MFCD01320232
  • Cas Number : 898751-96-7
  • Molecular Weight : 331.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7538F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7538F
Availability details for
Quantity
  • Availability
  • Add to Cart