RIE156330256 Rieke Metals, Inc.

Ethyl 4-(3-methoxyphenyl)-4-oxobutyrate

  • MDL number MFCD01311609
  • Cas Number : 147373-96-4
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7540B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7540B
Availability details for
Quantity
  • Availability
  • Add to Cart