RIE156330257 Rieke Metals, Inc.

Ethyl 5-(3-methoxyphenyl)-5-oxovalerate

  • MDL number MFCD01311612
  • Cas Number : 898751-99-0
  • Molecular Weight : 250.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7540C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7540C
Availability details for
Quantity
  • Availability
  • Add to Cart