RIE156330258 Rieke Metals, Inc.

Ethyl 6-(3-methoxyphenyl)-6-oxohexanoate

  • MDL number MFCD01311611
  • Cas Number : 898752-02-8
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7540D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7540D
Availability details for
Quantity
  • Availability
  • Add to Cart