RIE156330260 Rieke Metals, Inc.

Ethyl 8-(3-methoxyphenyl)-8-oxooctanoate

  • MDL number MFCD01320233
  • Cas Number : 898752-05-1
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7540F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7540F
Availability details for
Quantity
  • Availability
  • Add to Cart